Learn More
Medchemexpress LLC Octanoic acid, 8-bromo-, 1-octylnonyl ester | 2089253-22-3 | MFCD34178802 | 97.0% | 461.56 | C25H49BrO2 | 500 MG

Supplier: Medchemexpress LLC HYW440813500MG
Heptadecan-9-yl 8-bromooctanoate is a brominated fatty ester used as a hydrophobic lipid component for building or modifying lipid nanoparticles. Supplied for research use only, it provides a halogenated alkyl ester functionality useful in formulation and lipid chemistry workflows.
- Brominated fatty ester useful for building or modifying lipid nanoparticles.
- Purity 97.0%.
- Molecular formula C25H49BrO2.
- Molecular weight 461.56.
- Available as a solid and as a 10 mM solution in DMSO for ready reconstitution.
- Suitable for formulation and lipid chemistry research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.