Learn More
Sigma Aldrich Fine Chemicals Biosciences Octanoic acid 99500ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C2875500ML
Encompass
Octanoic acid is an eight carbon fatty acid which is found in palm coconut oil and milk. It is used to treat candidiasis and bacterial infections. Octanoic acid has antibacterial and antifungal properties. It is used in nutritional supplementation.
Catalog No. 50-290-3785