Learn More
Apexbio Technology LLC (±)-Octanoylcarnitine chloride 14919-35-8 25mg
SDP
Supplier:  Apexbio Technology LLC B637125
Encompass_Preferred
( )-Octanoylcarnitine chloride is an acyl derivative structurally related to acetylcarnitine chloride it functions as an intermediate in fatty acid metabolism by facilitating fatty acid transport across mitochondrial membranes for subsequent -oxidation and ATP synthesis Experimental evidence indicates that ( )-octanoylcarnitine modulates mitochondrial utilization of certain branched-chain 2-oxo acids influencing their oxidation rates in isolated mitochondria Additionally researchers employ ( )-octanoylcarnitine combined with malate to investigate mitochondrial respiratory activity examining oxidative phosphorylation dynamics in response to metabolic modulators such as ADP and creatine particularly in isolated cardiac muscle tissues
Catalog No. 50-000-63741