Learn More
Medchemexpress LLC Octenidine dihydrochloride | 70775-75-6 | 99.7% | 1 G

Supplier: Medchemexpress LLC HYB2170A1G
Octenidine dihydrochloride is an effective antiseptic compound for skin, mucous membranes, and wounds. It is for research use only.
- Antiseptic compound: effective for skin, mucous membranes, and wounds.
- Microbicidal activity: at concentrations less than 1.5 μM (0.94 μg/mL), it causes a greater than 99% reduction of tested microbial populations within 15 minutes.
- Broad spectrum: effective against Staphylococcus epidermidis, E. coli, and C. albicans.
- In vivo efficacy: maintains antimicrobial activity when applied to the skin of hands and feet.
- Dental plaque reduction: daily topical application of 1% solutions significantly reduces plaque score on buccal tooth surfaces.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.