Learn More
Sartorius Sartorius Octet 10mM Glycine Regeneration Buffer, Enables Biosensor Re-Use, Cost-Effective for High-Throughput Analysis, 100 mL
Supplier:  Sartorius 181184
Encompass
The Sartorius Octet 10mM Glycine pH 1.7 is a specialized regeneration buffer designed for Octet BLI biosensors. It facilitates efficient dissociation of bound analytes, enabling re-use of biosensors for high-throughput analysis. This cost-effective solution supports various applications in kinetics and quantitation assays, making it ideal for laboratories conducting extensive analyses with multiple samples or ligand-analyte pairs.
Catalog No. NC3234293