Learn More
Sartorius OCTET-AS SYSTEM
Supplier:  Sartorius OCTETAS
Encompass
The Octet AS Offline Biosensor Immobilization Station is designed for use with Octet BLI systems to reduce assay times while improving the efficiency of the assay. Octet AS allows the users to process assay steps that do not require online signal monitoring such as biosensor loading inter-step buffer washers or any intermediate assay steps freeing up the instrument time.
Catalog No. 50-289-1772
Request a Quote
To request a quote for this item, please select Request a Quote to reach our dedicated support team.