Learn More
Sartorius Sartorius Octet Protein A Biosensors, Rapid Quantitation, High Throughput, Tray
Supplier:  Sartorius 185010
Encompass
Sartorius Octet Protein A Biosensors are designed for rapid and precise quantitation of immunoglobulins, including human IgG, using advanced bioprocessing applications. Their unique Protein A coating allows direct measurement from complex samples, offering speed and accuracy with a throughput of ~32 min for 96 samples. The regeneration feature and high-throughput capabilities make them ideal for critical tasks like protein purification and cell line screening.
Catalog No. NC2939019