Learn More
Sartorius Octet SPR MAR Sensor Chip
Supplier:  Sartorius 190115
Encompass
The Octet SPR MAR sensor chip is suitable for a wide range of SPR assays due to its versatility and high binding capacity – from small fragment research to large complex molecular structures. The Octet SPR MAR sensor chip is comprised of a 3D carboxymethylated dextran matrix sensor chip surface which can covalently bond to molecules of interest through a range of different amine aldehyde thiol or carboxyl functional groups.
Catalog No. 50-289-1683