Learn More
Sartorius Sartorius Octet AT Biosensor Transfer Tool, Ergonomic Design, Transfers 8 Biosensors, Research Use Only
Supplier:  Sartorius 185159
Encompass
The Sartorius Octet AT Biosensor Transfer Tool is an ergonomic device designed to enhance efficiency in biosensor transfer for Bio-Layer Interferometry (BLI). It allows the simultaneous transfer of eight biosensors, featuring a unique finger hook for improved stability and reduced user fatigue. This lightweight tool is compatible with the Sartorius Linear Pipette Stand, making workflows quicker and more efficient in research and bioprocessing environments.
Catalog No. NC3314746