Learn More
Cayman Chemical OctnoIc AcId-d2 10mg
SDP
Supplier:  Cayman Chemical 2941210 MG
Encompass_Preferred
An internal standard for the quantification of octanoic acid by GC- or LC-MS
Catalog No. 50-298-9557