Learn More
Cayman Chemical OctnoIc AcId-d2 5mg
SDP
Supplier:  Cayman Chemical 294125 MG
Encompass_Preferred
An internal standard for the quantification of octanoic acid by GC- or LC-MS
Catalog No. 50-298-9298