Learn More
Cayman Chemical OctnoIc AcId methyl ester 100g
SDP
Supplier:  Cayman Chemical 26717100 G
Encompass_Preferred
A fatty acid methyl ester; has been found in biodiesels made from the transesterification of beef tallow, soybean oil, and babassu oil blends; an aromatic volatile compound in cantaloupe, galia, and honeydew melons
Catalog No. 50-293-4729