Learn More
Medchemexpress LLC Octocrylene | 6197-30-4 | 98.7% | C24H27NO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYA0087500MG
Encompass_Preferred
Octocrylene is an organic ultraviolet (UV) filter known for its ability to absorb UVB radiation and shorter UVA wavelengths. It functions as a partial agonist of PPARγ, influencing the gene transcription profile of lipid metabolism enzymes. Research also indicates its involvement in cellular processes like cytotoxicity and estrogen biosynthesis.
  • Acts as an organic ultraviolet (UV) filter
  • Primarily absorbs UVB and shorter UVA wavelengths
  • Functions as a partial agonist of PPARγ
  • Influences gene transcription of lipid metabolism enzymes
  • Exhibits cytotoxicity and genotoxicity in human skin fibroblasts
  • Mediates estrogen biosynthesis in zebrafish larvae
  • Impacts antioxidant pathways including glutathione transferase
Catalog No. 50-004-32108