Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octocrylene (standard) | 6197-30-4 | 98.1% | C₂₄H₂₇NO₂ | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYA0087R

Encompass_Preferred

Octocrylene (Standard) is an analytical standard for research and analytical applications, including HPLC, GC, and MS. It acts as an organic ultraviolet (UV) filter, absorbing primarily UVB radiation and short UVA wavelengths. This makes it suitable for cosmetic products, providing sun protection in sunscreens or protecting formulations from UV radiation. It appears as a light yellow to yellow oil, with a molecular weight of 361.48 and a chemical formula of C₂₄H₂₇NO₂.

  • Analytical standard: serves as a reference.
  • UV filter: absorbs mainly UVB and short UVA wavelengths.
  • Cosmetic application: suitable for sunscreens and protecting formulations.
  • Research applications: used in HPLC, GC, MS.
  • Appearance: light yellow to yellow oil.
  • High purity: 98.08%.

Catalog No. 50-003-58584


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.