Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Octodrine hydrochloride | 5984-59-8 | 98.0% | 1 G
SDP

Supplier:  Medchemexpress LLC HYB1233A1G

Encompass_Preferred

Octodrine (2-Amino-6-methylheptane) is identified as a central nervous activator. It functions by increasing the uptake of dopamine and noradrenaline. Studies have indicated that Octodrine can increase the pain threshold, cardiac rate (exhibiting a positive chronotropic effect), and myocardial contractility (showing a positive inotropic effect). This product is intended for research use only and is not sold to patients.

  • Increases uptake of dopamine and noradrenaline.
  • Increases pain threshold.
  • Increases cardiac rate (positive chronotropic effect).
  • Increases myocardial contractility (positive inotropic effect).
  • For research use only.
  • Store at 4°C, sealed, away from moisture.

Catalog No. 50-004-12993


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.