Learn More
Medchemexpress LLC Octodrine hydrochloride | 5984-59-8 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB1233A5G
Encompass_Preferred
Octodrine (2-Amino-6-methylheptane) is a central nervous activator that increases the uptake of dopamine and noradrenaline. It is found to increase the pain threshold, cardiac rate (positive chronotropic effect), and myocardial contractility (positive inotropic effect).
  • Increases the uptake of dopamine and noradrenaline
  • Increases pain threshold
  • Increases cardiac rate (positive chronotropic effect)
  • Increases myocardial contractility (positive inotropic effect)
  • Functions as a central nervous activator
Catalog No. 50-004-12872