Learn More
Apexbio Technology LLC (+,-)-Octopamine HCl 770-05-8 5g
SDP
Supplier:  Apexbio Technology LLC B19955000
Encompass_Preferred
( -)-Octopamine HCl (CAS 770-05-8) is a biogenic monoamine structurally analogous to noradrenaline In invertebrates it functions as a neurohormone neuromodulator and neurotransmitter by interacting with specific octopamine receptors to regulate physiological and behavioral processes including metabolism locomotion and learning The compound is widely utilized in neurobiological research to investigate aminergic signaling pathways synaptic transmission and the functional roles of monoamines in invertebrate nervous systems Its use facilitates studies on neurotransmitter mechanisms distinct from those predominant in vertebrates
Catalog No. 50-000-93210