Learn More
Cayman Chemical OctopInIc AcId 100ug
SDP
Supplier:  Cayman Chemical 33093100 UG
Encompass_Preferred
An opine; has been found in crown gall tumors induced by the plant pathogenic bacterium A. tumefaciens
Catalog No. 50-294-7011