Learn More
Carbosynth LLC. Octreotide, 1mg, Somatostatin Analog, Disulfide bonds: Cys2-Cys7, MW: 1019.24, Molecular formula: C49H66N10O10S2, CAS# 83150-76-9
SDP
Supplier:  Carbosynth LLC. OCT3738PI5MG
Encompass_Preferred
Synonyms: D-Phe-(Cys-Phe-D-Trp-Lys-Thr-Cys)-Thr-ol, Salt Form:, Trifluoroacetate, Storage Temperature: -20°C
Catalog No. 50-168-6296