Learn More
Bachem Octreotide Acetate
SDP
Supplier:  Bachem 4042822.0005
Encompass_Preferred
Octreotide Acetate, H-D-Phe-Cys-Phe-D-Trp-Lys-Thr-Cys-L-threoninol, 79517-01-4, C49H66N10O10S2, Mr 1019.26, 5mg. (D-Phe5,Cys6·11,D-Trp8,L-Threoninol12)-Somatostatin-14 (5-12), SMS 201-995. Octreotide is a longer acting synthetic octapeptide analog of the naturally occurring hormone somatostatin. It inhibits the secretion of gastro-entero-pancreatic peptide hormones and the release of growth hormones. Melacini et al. studied the conformation of the peptide in solution by NMR. CAS Number (net): 83150-76-9.
Catalog No. 50-260-015