Learn More
Apexbio Technology LLC Octreotide acetate, 83150-76-9, MFCD08277638, 50 mg
SDP
Supplier:  Apexbio Technology LLC B497950
Encompass_Preferred
MF: C51H70N10O12S2, MW: 1079.29, Synonyms: SMS 201995; Sandostatin. Solubility: 53.9645mg/mL in DMSO. Octreotide acetate (Sandostatin) is an octapeptide congener of native somatostatin, inhibits the secretion of insulin and glucagon. It reduces production of IGF-1 and IGF-2 by the liver by modulation of growth-hormone secretion from the pituitary gland.
Catalog No. 50-115-2086