Learn More
Selleck Chemical LLC Octreotide Acetate 50mg 79517-01-4 D-Phe-c[Cys-Phe-D-Trp-Lys-Thr-Cys]-Thr-ol
SDP
Supplier:  Selleck Chemical LLC P101750MG
Encompass_Preferred
Octreotide Acetate is the acetate salt of a cyclic octapeptide. It is a long-acting octapeptide with pharmacologic properties mimicking those of the natural hormone somatostatin. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8069