Learn More
American Radiolabeled Chemicals Inc OCTYL 14C B GLUCOPYR 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 363450UC
Encompass_Preferred
Octyl-1-14C b-D-glucopyranoside 50uCi in ethanol:water (7:3), 0.1 mCi/ml, 10-20 mCi/mmol, M.W. 292.38, Exclusive
Catalog No. 50-352-966
Add to Cart
Request a Quote