Learn More
Medchemexpress LLC Octyl-a-ketoglutarat 1mg | 876150-14-0 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1143331MG
Encompass_Preferred
Octyl-α-ketoglutarate is a cell-permeable ester derivative of α-ketoglutarate used in biochemical and cell biology research to increase intracellular α-ketoglutarate and modulate α-ketoglutarate-dependent enzymes such as prolyl hydroxylases. Molecular formula C13H22O5; molecular weight 258.31 g/mol. Supplied as a solid powder; follow the supplier datasheet for storage and handling.
  • Cell-permeable α-ketoglutarate ester.
  • Increases intracellular α-ketoglutarate levels.
  • Used to study HIF regulation and prolyl hydroxylase activity.
  • Supplied as a solid; follow the datasheet for storage and handling.
Catalog No. 50-002-29679