Learn More
Medchemexpress LLC Octyl gallate | 1034-01-1 | 282.33 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN20111G
Encompass_Preferred
Octyl gallate (Progallin O) is extensively utilized as a food additive due to its antimicrobial and antioxidant properties. It exhibits a selective and sensitive fluorescent characteristic and demonstrates a significant antiviral effect against HSV-1, vesicular stomatitis virus (VSV), and poliovirus.
  • Food additive
  • Antimicrobial activity
  • Antioxidant activity
  • Selective and sensitive fluorescent property
  • Antiviral effect against HSV-1, vesicular stomatitis virus (VSV), and poliovirus
Catalog No. 50-004-15920