Learn More
Medchemexpress LLC Octyl gallate | 1034-01-1 | 282.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN2011R100MG
Encompass_Preferred
Octyl gallate (Progallin O) is widely used as a food additive with antimicrobial and antioxidant activity. It also shows selective and sensitive fluorescent properties and has a marked antiviral effect against HSV-1, vesicular stomatitis virus (VSV), and poliovirus. It also exhibits antimicrobial activity over H. pylori, with a minimum inhibitory concentration (MIC) value of 125 μg/mL.
  • Widely used as a food additive.
  • Exhibits antimicrobial and antioxidant activity.
  • Shows selective and sensitive fluorescent properties.
  • Possesses marked antiviral effect against HSV-1, vesicular stomatitis virus (VSV), and poliovirus.
  • Demonstrates antimicrobial activity over H. pylori, with a minimum inhibitory concentration (MIC) value of 125 μg/mL.
Catalog No. 50-004-15164