Learn More
Medchemexpress LLC Octyl gallate | 1034-01-1 | 282.33 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN20115G
Encompass_Preferred
Octyl gallate (Progallin O) is widely used as a food additive, possessing antimicrobial and antioxidant activity. It also exhibits selective and sensitive fluorescent properties. Furthermore, Octyl gallate has demonstrated a marked antiviral effect against HSV-1, vesicular stomatitis virus (VSV), and poliovirus.
  • Widely used as a food additive
  • Possesses antimicrobial and antioxidant activity
  • Exhibits selective and sensitive fluorescent properties
  • Demonstrates a marked antiviral effect against HSV-1, vesicular stomatitis virus (VSV), and poliovirus
Catalog No. 50-004-16275