Learn More
Medchemexpress LLC Odanacatib | 603139-19-1 | 99.8% | 525.56 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY10042
Encompass_Preferred
Odanacatib (MK-0822) is a potent and selective inhibitor of cathepsin K, with an IC50 of 0.2 nM for human cathepsin K. It is a weak inhibitor of antigen presentation and reduces resorption activity.
  • Potent and selective inhibitor of cathepsin K
  • Weak inhibitor of antigen presentation
  • Reduces resorption activity
  • Suppresses bone resorption and serum bone formation markers
  • Increases periosteal bone formation and cortical thickness
  • Promotes osseointegration of titanium implants
Catalog No. 50-003-47194