Learn More
Medchemexpress LLC Odetiglucan | 53238-80-5 | 16 kDa | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147295100MG
Encompass_Preferred
Odetiglucan is a novel β-glucan that functions as a potent immunostimulant and a Dectin-1 (CLEC7A) agonist. It activates innate immune effector cells and triggers a coordinated anti-cancer immune response.
  • Novel β-glucan compound
  • Potent immunostimulant
  • Dectin-1 (CLEC7A) agonist
  • Activates innate immune effector cells
  • Triggers a coordinated anti-cancer immune response
Catalog No. 50-003-60640