Learn More
Medchemexpress LLC Odm-203 | 1430723-35-5 | 99.6% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11936750MG
Encompass_Preferred
ODM-203 is an orally active small-molecule inhibitor that targets FGFR1-4 and VEGFR1-4, provided for research use only. Clinical development was conducted by Orion Pharma; the listing reflects research-grade material for preclinical studies.
  • Dual FGFR and VEGFR inhibition with low-nanomolar IC50 values.
  • High purity research-grade material (~99.6%).
  • Available as powder and as 10 mM solution in DMSO.
  • Multiple solid pack sizes, including 50 MG.
  • Intended for in vitro and in vivo preclinical research; not for human use.
Catalog No. 50-002-33641