Learn More
Medchemexpress LLC HY-119367 5mg Medchemexpress, ODM-203 CAS:1430723-35-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1193675MG
Encompass_Preferred
Medchemexpress, HY-119367 5mg ODM-203 CAS:1430723-35-5 ODM-203 is a potent FGFR and VEGFR families inhibitor with IC50s of 11, 16, 6, 35 nM towards recombinant FGFR1, FGFR2, FGFR3 and FGFR4 as well as 26, 9, 5 nM towards VEGFR1, VEGFR2 and VEGFR3, respectively. ODM-203 exhibits strong anti-tumor activity and induces anti-tumor immunity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9530