Learn More
Medchemexpress LLC ODM-207 1mg | 1801503-93-4 | 99.6% | 375.42 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1119161MG
Encompass_Preferred
ODM-207, also known as BET-IN-4, is a highly pure compound that functions as a potent inhibitor of the BET bromodomain protein BRD4, with an IC50 of ≤ 1 μM. It is suitable for various in vitro and in vivo studies.
  • Potent BRD4 inhibitor
  • IC50 of ≤ 1 μM
  • High purity
  • Off-white to light yellow solid appearance
  • Multiple solubility protocols available for research
Catalog No. 50-004-39350