Learn More
Medchemexpress LLC ODN 1585 | 386832-46-8 | DNA, d(G-sp-G-G-G-T-C-A-A-C-G-T-T-G-A-G-G-sp-G-sp-G-sp-G-sp-G) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1507255MG
Encompass_Preferred
ODN 1585 is a potent inducer of IFN and TNFα production. It is also a potent stimulator of NK (natural killer) function and increases CD8+ T-cell function, including the CD8+ T cell-mediated production of IFN-γ. It induces regression of established melanomas in mice and can confer complete protection against malaria in mice.
  • Potent inducer of IFN and TNFα production.
  • Stimulates NK (natural killer) cell function.
  • Increases CD8+ T-cell function.
  • Induces regression of established melanomas in mice.
  • Confers complete protection against malaria in mice.
  • Used for acute myelogenous leukemia (AML) research.
  • Used for malaria research.
  • Functions as a vaccine adjuvant.
Catalog No. 50-003-75663