Learn More
Medchemexpress LLC 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine N-hydroxysuccinimide ester | 1010188-79-0 | 90.0% | 955.21 | C50H87N2O13P | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15024110MG
Encompass_Preferred
DOPE-NHS is an NHS-ester functionalized phospholipid used as a membrane-anchored linker to covalently attach peptides and other primary-amine-containing ligands to exosomes, liposomes, and membrane-based nanoparticles. It is intended for research applications such as surface modification, targeting, and drug-delivery formulation and is provided as an oily material with documented solubility in DMSO.
Catalog No. 50-002-87151