Learn More
Medchemexpress LLC ODN 1826 sodium | 00-00-0 | 99.5% | 6364.1 (free acid) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY146245C5MG
Encompass_Preferred
ODN 1826 sodium is a class B CpG oligodeoxynucleotide that functions as a TLR9 agonist for research applications. It stimulates innate immune responses, inducing nitric oxide (NO) and iNOS production, and can enhance apoptosis and antitumor immune surveillance in preclinical studies. Supplied as a sodium salt with a defined phosphorothioate-modified sequence and high purity, it is intended for research use only.
  • Acts as a TLR9 agonist to stimulate innate immunity.
  • Induces nitric oxide and iNOS production.
  • Enhances apoptosis in responsive cells.
  • Shows reported antitumor activity in preclinical models.
  • Provided as a sodium salt, solid, white to off-white.
  • High purity (99.5%).
  • Store dry, sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • For research use only.
Catalog No. 50-002-22745