Learn More
Medchemexpress LLC ODN 2216 | 332437-00-0 | DNA, d(G-sp-G-sp-G-G-G-A-C-G-A-T-C-G-T-C-G-sp-G-sp-G-sp-G-sp-G-sp-G) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1507415MG
Encompass_Preferred
ODN 2216 is a human-specific Toll-like Receptor 9 (TLR9) ligand or agonist. It is known to induce high amounts of IFN-α and IFN-β, and IFN-α by plasmacytoid DC (pDC) and IL-12 (p40) production by dendritic cells (DC). It also stimulates IFN-γ production in peripheral blood mononuclear cells (PBMC), which is an indirect effect mediated by IFN-α/β. ODN 2216 can activate NK cells and promote IFN-γ production of TCR-triggered CD4+ T cells and is intended for research use only.
  • Induces high amounts of IFN-α and IFN-β
  • Stimulates IFN-γ production in peripheral blood mononuclear cells
  • Activates NK cells
  • Promotes IFN-γ production of TCR-triggered CD4+ T cells
Catalog No. 50-003-75854