Learn More
Medchemexpress LLC Odn 2336 (sodium) | DNA, d(G-sp-G-sp-G-G-A-C-G-A-C-G-T-C-G-T-G-G-sp-G-sp-G-sp-G-sp-G), sodium salt | 1 MG
SDP
Supplier:  Medchemexpress LLC HY150742A1MG
Encompass_Preferred
ODN 2336 (sodium) is a laboratory chemical provided as a solid. It is stable under recommended storage conditions and is not classified as a hazardous substance or mixture.
  • Recommended for laboratory use and substance manufacturing
  • Stable when stored under recommended conditions
  • Store sealed at 4°C, away from moisture
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
  • Ships at room temperature for durations less than two weeks
  • Use in well-ventilated areas with appropriate exhaust systems
Catalog No. 50-003-74989