Learn More
Medchemexpress LLC ODN 24888 sodium | 1964506-30-6 | DNA, d(P-thio)(T-C-C-T-G-G-C-c7rGm-G-G-G-A-A-G-T) | 1 MG
SDP
Supplier:  Medchemexpress LLC HY150744A1MG
Encompass_Preferred
ODN 24888 sodium is a guanine-modified inhibitory oligonucleotide (INH-ODN) that potently inhibits TLR7/TLR9-mediated signaling. It impairs IFN-α levels and NF-κB activation, and inhibits IL-6 release. This product is involved in immune and inflammatory responses and can be utilized as a vaccine adjuvant.
  • Guanine-modified inhibitory oligonucleotide
  • Potent inhibition of TLR7/TLR9-mediated signaling
  • Impairs IFN-α levels and NF-κB activation
  • Inhibits IL-6 release
  • Involved in immune and inflammatory responses
  • Can be used as a vaccine adjuvant
Catalog No. 50-003-74924