Learn More
Medchemexpress LLC ODN 6016 sodium | DNA, d(T-sp-C-G-A-C-G-T-C-G-T-G-G-sp-G-sp-G-sp-G), sodium salt | 1 MG
Supplier: Medchemexpress LLC HY150747A1MG
ODN 6016 sodium is classified as a CpG-A oligonucleotide. This compound is known for its ability to induce the production of IFN-α. Researchers utilize ODN 6016 sodium in studies concerning immune disorders, particularly those involving immunodeficiency, such as conditions caused by HIV-1.
- Type: CpG-A oligonucleotide
- Function: Induces IFN-α production
- Research applications: Immune disorders, immunodeficiency (e.g., HIV-1-related)
- Sequence: T-sp-C-G-A-C-G-T-C-G-T-G-G-sp-G-sp-G-sp-G