Learn More
YMC America, Inc ODS-A 120A 3um 5x0.5mm 1/32
SDP
Supplier:  YMC America, Inc AA12S03E5J0RU
Encompass
YMC-Pack ODS-A is a high-coverage fully-endcapped C18 phase which exhibits exceptional lot-to-lot reproducibility It is available in three different pore sizes and is suitable for the separation of a wide range of compounds YMC-Pack ODS-A in 120 pore size is used for high performance reversed phase separations It has selectivity for the separation of many organic compounds found in pharmaceutical chemical and biotechnology industries 200 ODS-A offers a unique mix of moderately large pores and high surface area for the analysis of compounds ranging from small organics and pharmaceuticals to moderately-sized proteins
Catalog No. 50-008-77201