Learn More
YMC America, Inc ODS-AL 120A 5um 150x0.5mm 1/16
SDP
Supplier:  YMC America, Inc AL12S0515J0AU
Encompass
YMC-Pack ODS-AL is a non-endcapped high carbon load monomeric C18 phase It exhibits different selectivity from conventional ODS columns due to the use of not only hydrophobic interaction but also secondary interactions caused by silanol groups YMC-Pack ODS-AL is intended for use only in those situations where residual silanol activity is beneficial to the separation
Catalog No. 50-008-78566