Learn More
YMC America, Inc ODS-AM 120A 3um 100x0.3mm 1/16
SDP
Supplier:  YMC America, Inc AM12S0310H0AU
Encompass
YMC-Pack ODS-AM is produced with specific emphasis on lot-to-lot reproducibility of the packing material Strict manufacturing control is performed on all aspects of quality including physical properties of the silica base gel as well as surface modifying processes ensuring exceptional reproducibility
Catalog No. 50-008-79544