Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YMC America, Inc ODS-AQ 50UM 1200A 500X50MM
SDP

Supplier:  YMC America, Inc AQ12S505051DR

Encompass

Ods-Aq 50Um; 1200A 500 X 50mm

Catalog No. 50-874-269


May include imposed supplier surcharges.
Only null left
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.