Learn More
LICOR Odyssey 28 KD Loading Indicator 700 nm (2 x 1 mL)
Supplier:  LICOR 92620004
Encompass_Preferred
The Odyssey 28 KD Loading Indicator - 700 nm is a single 28 kDa recombinant protein directly labeled with a near-infrared fluorescent dye. You can detect the 700 nm version in the 700 nm channel of your imaging system.
Catalog No. NC1551074