Learn More
AdipoGen Oenanthacid-4-CF3-umbellifer
SDP
Supplier:  AdipoGen CDXO0006M050
Encompass_Preferred
Chemical. CAS 351002-79-4. Formula C17H17F3O4. MW 342.31. Synthetic. Suitable as fluorogenic substrate for lipases.
Catalog No. 50-256-0355