Learn More
TARGETMOL CHEMICALS INC OF-1 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T212710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. OF-1 is a potent inhibitor of BRPF1B and BRPF2 bromodomain. purity: 98%
Catalog No. 50-257-7220