Learn More
Selleck Chemical LLC OF-1 5mg 919973-83-4
SDP
Supplier:  Selleck Chemical LLC S76815MG
Encompass_Preferred
OF-1 is a potent inhibitor of BRPF1B and BRPF2 bromodomain with Kd of 100 nM and 500 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5119