Learn More
Med Vet International Ofloxacin Ophthalmic Solution, 0.3%, 5mL
Supplier:  Med Vet International RX24208043405
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Ofloxacin ophthalmic solution is used to treat bacterial infections of the eye, including conjunctivitis (pink eye) and ulcers of the cornea. Ofloxacin is in a class of medications called quinolone antibiotics. It works by killing bacterial cells that cause infection.
Catalog No. 50-282-8408