Learn More
Medchemexpress LLC Ogerin | 1309198-71-7 | 99.9% | 307.35 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11027910MM/1ML
Encompass_Preferred
Ogerin is a chemical probe and a selective GPR68 positive allosteric modulator (PAM) with a pEC50 of 6.83. It also has a moderate antagonistic effect on A2A (Ki=220 nM). Ogerin inhibits the fear conditioning reflex in mice and prevents TGF-β-induced myofibroblast differentiation of fibroblasts from multiple organ systems. It is suitable for studies related to fibrotic diseases and neurological disorders.
  • Selective GPR68 positive allosteric modulator (PAM).
  • Moderate antagonistic effect on A2A (Ki=220 nM).
  • Inhibits fear conditioning reflex in mice.
  • Prevents TGF-β-induced myofibroblast differentiation of fibroblasts.
  • Suitable for studies related to fibrotic diseases.
  • Suitable for studies related to neurological disorders.
Catalog No. 50-003-44104