Learn More
Medchemexpress LLC HY-100898 5mg Medchemexpress, OGT 2115 CAS:853929-59-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1008985MG
Encompass_Preferred
Medchemexpress, HY-100898 5mg OGT 2115 CAS:853929-59-6 OGT 2115 is a potent, cell-permeable and orally active heparanase inhibitor with an IC50 of 0.4 μM. OGT 2115 has anti-angiogenic properties (IC50 of 1 μM). OGT 2115 also inhibits heparan sulfate degradation activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7790